2-{[(4-Chlorophenyl)sulfonyl]amino}-2-phenylacetic acid ≥95%

Supplier: Matrix Scientific

042539-500MG 042539-1G 042539-5G
101856-248EA 181.54 USD
101856-248 101856-250 101856-252
2-{[(4-Chlorophenyl)sulfonyl]amino}-2-phenylacetic acid ≥95%
2-{[(4-Chlorophenyl)sulfonyl]amino}-2-phenylacetic acid

Hazard: Irritant

Formula: C₁₄H₁₂ClNO₄S
MW: 325.77 g/mol
Melting Pt: 175…177 °C
MDL Number: MFCD00172800
CAS Number: 117309-47-4

Order Now

Specification Test Results

Melting point 175-177°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR