3-{[3-(Trifluoromethyl)benzyl]sulfanyl}-2-thiophenecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-[3-(Trifluoromethyl)benzylthio]thiophene-2-carboxylic acid

042538-500MG 042538-1G
101856-244EA 556.92 USD
101856-244 101856-246
3-{[3-(Trifluoromethyl)benzyl]sulfanyl}-2-thiophenecarboxylic acid ≥95%
3-{[3-(Trifluoromethyl)benzyl]sulfanyl}-2-thiophenecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₉F₃O₂S₂
MW: 318.34 g/mol
Melting Pt: 178…180 °C
MDL Number: MFCD00663855
CAS Number: 251096-78-3

Order Now

Specification Test Results

Melting point 178-180°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR