1-[(Ammoniooxy)methyl]-3-chlorobenzene chloride ≥95%

Supplier: Matrix Scientific

Synonyms: O-(3-Chlorobenzyl)hydroxylamine hydrochloride

042532-500MG 042532-1G 042532-5G
101856-220EA 164.62 USD
101856-220 101856-222 101856-224
1-[(Ammoniooxy)methyl]-3-chlorobenzene chloride ≥95%
1-[(Ammoniooxy)methyl]-3-chlorobenzene chloride

Hazard: Irritant

Formula: C₇H₉Cl₂NO
Melting Pt: 206-212°C
MDL Number: MFCD01114569
CAS Number: 29605-78-5

Order Now

Specification Test Results

Melting point 206-212°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR