2-{[5-(1-Benzyl-2-oxo-1,2-dihydro-3-pyridinyl)-4-methyl-4H-1,2,4-triazol-3-yl]sulfanyl}propanoic acid ≥95%

Supplier: Matrix Scientific

042526-500MG 042526-1G 042526-5G
101856-190EA 921.54 USD
101856-190 101856-192 101856-194
2-{[5-(1-Benzyl-2-oxo-1,2-dihydro-3-pyridinyl)-4-methyl-4H-1,2,4-triazol-3-yl]sulfanyl}propanoic acid ≥95%
2-{[5-(1-Benzyl-2-oxo-1,2-dihydro-3-pyridinyl)-4-methyl-4H-1,2,4-triazol-3-yl]sulfanyl}propanoic acid

Hazard: Irritant

Formula: C₁₈H₁₈N₄O₃S
Melting Pt: 194…197 °C
MDL Number: MFCD00141403
CAS Number: 282523-27-7

Order Now

Specification Test Results

Melting point 194-197°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR