2-{[5-(1-Benzyl-2-oxo-1,2-dihydro-3-pyridinyl)-4-methyl-4H-1,2,4-triazol-3-yl]sulfanyl}acetic acid ≥95%

Supplier: Matrix Scientific

042525-500MG 042525-1G 042525-5G
101856-184EA 921.54 USD
101856-184 101856-186 101856-188
2-{[5-(1-Benzyl-2-oxo-1,2-dihydro-3-pyridinyl)-4-methyl-4H-1,2,4-triazol-3-yl]sulfanyl}acetic acid ≥95%
2-{[5-(1-Benzyl-2-oxo-1,2-dihydro-3-pyridinyl)-4-methyl-4H-1,2,4-triazol-3-yl]sulfanyl}acetic acid

Hazard: Irritant

Formula: C₁₇H₁₆N₄O₃S
Melting Pt: 190…192 °C
MDL Number: MFCD00141402
CAS Number: 282523-25-5

Order Now

Specification Test Results

Melting point 190-192°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR