Ethyl-2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}acetate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl N-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]glycinate

042522-1G 042522-5G
101856-170EA 240 USD
101856-170 101856-172
Ethyl-2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}acetate ≥95%
Ethyl-2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}acetate

Hazard: Irritant

Formula: C₁₀H₁₀ClF₃N₂O₂
MW: 282.65 g/mol
Melting Pt: 78…80 °C
MDL Number: MFCD00141296
CAS Number: 246022-36-6

Order Now

Specification Test Results

Melting point 78-80°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR