1-Benzyl-3-(4-methyl-5-sulfanyl-4H-1,2,4-triazol-3-yl)-2(1H)-pyridinone ≥95%

Supplier: Matrix Scientific

042519-500MG 042519-1G 042519-5G
101856-158EA 747.69 USD
101856-158 101856-160 101856-162
1-Benzyl-3-(4-methyl-5-sulfanyl-4H-1,2,4-triazol-3-yl)-2(1H)-pyridinone ≥95%
1-Benzyl-3-(4-methyl-5-sulfanyl-4H-1,2,4-triazol-3-yl)-2(1H)-pyridinone

Hazard: Irritant

Formula: C₁₅H₁₄N₄OS
Melting Pt: 253…255 °C
MDL Number: MFCD00140473
CAS Number: 242472-19-1

Order Now

Specification Test Results

Melting point 253-255°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR