1-Benzyl-2-oxo-1,2-dihydro-3-pyridinecarbohydrazide ≥95%
Supplier: Matrix Scientific
042518-500MG
042518-1G
042518-5G
101856-152EA
556.92
USD
101856-152
101856-154
101856-156
1-Benzyl-2-oxo-1,2-dihydro-3-pyridinecarbohydrazide ≥95%
1-Benzyl-2-oxo-1,2-dihydro-3-pyridinecarbohydrazide
Hazard: Irritant
Formula:
C₁₃H₁₃N₃O₂ Melting Pt: 110…114 °C |
MDL Number:
MFCD00140469 CAS Number: 242472-15-7 |
Specification Test Results
Melting point | 110-114°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...