5-Chloro-1-(2,4-dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

042517-500MG 042517-1G 042517-5G
101856-146EA 556.92 USD
101856-146 101856-148 101856-150
5-Chloro-1-(2,4-dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid ≥95%
5-Chloro-1-(2,4-dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid

Hazard: Irritant

Formula: C₁₃H₈Cl₃NO₃
Melting Pt: 258…260 °C
MDL Number: MFCD00140363
CAS Number: 242797-47-3

Order Now

Specification Test Results

Melting point 258-260°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR