1-(3,4-Difluorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

042516-500MG 042516-1G 042516-5G
101856-140EA 600 USD
101856-140 101856-142 101856-144
1-(3,4-Difluorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid ≥95%
1-(3,4-Difluorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid

Hazard: Irritant

Order Now

Specification Test Results

Melting point 214-216°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR