7-Chloro-2,4-bis(trifluoromethyl)[1,8]naphthyridine ≥95%

Supplier: Matrix Scientific

042508-500MG 042508-1G 042508-5G
101856-102EA 460 USD
101856-102 101856-104 101856-106
7-Chloro-2,4-bis(trifluoromethyl)[1,8]naphthyridine ≥95%
7-Chloro-2,4-bis(trifluoromethyl)[1,8]naphthyridine

Hazard: Irritant

Formula: C₁₀H₃ClF₆N₂
MW: 300.59 g/mol
Melting Pt: 92 °C
MDL Number: MFCD00140156
CAS Number: 106582-41-6

Order Now

Specification Test Results

Melting point 92°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR