3-(Dimethylamino)-1-(4-fluorophenyl)-2-propen-1-one ≥95%

Supplier: Matrix Scientific

042506-500MG 042506-1G 042506-5G
101856-090EA 181.54 USD
101856-090 101856-092 101856-094
3-(Dimethylamino)-1-(4-fluorophenyl)-2-propen-1-one ≥95%
3-(Dimethylamino)-1-(4-fluorophenyl)-2-propen-1-one

Hazard: Irritant

Formula: C₁₁H₁₂FNO
MW: 193.22 g/mol
Melting Pt: 84…86 °C
MDL Number: MFCD00111509
CAS Number: 75175-77-8

Order Now

Specification Test Results

Melting point 84-86°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR