Methyl-2-[2-(dimethylamino)-1-(methoxycarbonyl)vinyl]benzenecarboxylate ≥95%

Supplier: Matrix Scientific

042505-500MG 042505-1G 042505-5G
101856-084EA 375.38 USD
101856-084 101856-086 101856-088
Methyl-2-[2-(dimethylamino)-1-(methoxycarbonyl)vinyl]benzenecarboxylate ≥95%
Methyl-2-[2-(dimethylamino)-1-(methoxycarbonyl)vinyl]benzenecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₇NO₄
Melting Pt: 80…82 °C
MDL Number: MFCD08056598
CAS Number: 885950-29-8

Order Now

Specification Test Results

Melting point 80-82°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR