3-(Dimethylamino)-2-(3-thienyl)acrylonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
(2Z)-3-(Dimethylamino)-2-(thiophen-3-yl)prop-2-enenitrile
042504-500MG
042504-1G
042504-5G
101856-078EA
200
USD
101856-078
101856-080
101856-082
3-(Dimethylamino)-2-(3-thienyl)acrylonitrile ≥95%
3-(Dimethylamino)-2-(3-thienyl)acrylonitrile
Hazard: Irritant
Formula:
C₉H₁₀N₂S MW: 178.26 g/mol Melting Pt: 74…76 °C |
MDL Number:
MFCD00127051 CAS Number: 165404-39-7 |
Specification Test Results
Melting point | 74-76°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...