3-(Dimethylamino)-2-(2-thienyl)acrylonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: (2Z)-3-(Dimethylamino)-2-(thiophen-2-yl)prop-2-enenitrile

042503-500MG 042503-1G 042503-5G
101856-072EA 181.54 USD
101856-072 101856-074 101856-076
3-(Dimethylamino)-2-(2-thienyl)acrylonitrile ≥95%
3-(Dimethylamino)-2-(2-thienyl)acrylonitrile

Hazard: Irritant

Formula: C₉H₁₀N₂S
MW: 178.26 g/mol
Melting Pt: 62…64 °C
MDL Number: MFCD00138239
CAS Number: 165404-37-5

Order Now

Specification Test Results

Melting point 62-64°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR