4-(2-Pyridinyl)-2-pyrimidinamine ≥95%
Supplier: Matrix Scientific
042500-500MG
042500-1G
042500-5G
101856-054EA
287.69
USD
101856-054
101856-056
101856-058
4-(2-Pyridinyl)-2-pyrimidinamine ≥95%
4-(2-Pyridinyl)-2-pyrimidinamine
Hazard: Irritant
Formula:
C₉H₈N₄ MW: 172.19 g/mol Melting Pt: 137…139 °C |
MDL Number:
MFCD00138711 CAS Number: 66521-65-1 |
Specification Test Results
Melting point | 137-139°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...