2-(1,3,4-Oxadiazol-2-yl)phenol ≥95%

Supplier: Matrix Scientific

042498-500MG 042498-1G 042498-5G
101856-042EA 93.85 USD
101856-042 101856-044 101856-046
2-(1,3,4-Oxadiazol-2-yl)phenol ≥95%
2-(1,3,4-Oxadiazol-2-yl)phenol

Hazard: Irritant

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 109-110°C
MDL Number: MFCD00020713
CAS Number: 1008-65-7

Order Now

Specification Test Results

Melting point 109-110°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR