Fmoc-Inp-OH

Supplier: Matrix Scientific

Synonyms: 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-piperidinecarboxylic acid, 1-Fmoc-isonipecotic acid, 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]isonipecotic acid, Fmoc-isonipecotic acid, 1-Fmoc-4-piperidinecarboxylic acid, Fmoc-Isn-OH, 1-Fmoc-piperidine-4-carboxylic acid

042421-1G 042421-5G
101855-738EA 15.38 USD
101855-738 101855-740
Fmoc-Inp-OH
Fmoc-Inp-OH

Hazard: Irritant

Formula: C₂₁H₂₁NO₄
MW: 351.402 g/mol
Melting Pt: 172…173 °C
MDL Number: MFCD00273475
CAS Number: 148928-15-8

Order Now

Specification Test Results

Melting point 172-173

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR