4-Amino-3-hydroxypyridine hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Hydroxypyridin-4-amine hydrochloride, 4-Aminopyridin-3-ol hydrochloride
035764-250MG
035764-1G
101839-984EA
113.85
USD
101839-984
101839-986
4-Amino-3-hydroxypyridine hydrochloride ≥95%
4-Amino-3-hydroxypyridine hydrochloride
Hazard: Irritant
Formula:
C₅H₆N₂O·ClH MW: 146.575 g/mol Melting Pt: 151…152 °C |
MDL Number:
MFCD08729308 CAS Number: 1206679-69-7 |
Specification Test Results
Melting point | 151-152°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...