Azetidine hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: Azetidine HCl

035688-5G 035688-25G
101839-680EA 232.31 USD
101839-680 101839-682
Azetidine hydrochloride ≥97%
Azetidine hydrochloride

Hazard: Irritant/Hygroscopic

Formula: C₃H₈ClN
MW: 93.56 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00191762
CAS Number: 36520-39-5

Order Now

Specification Test Results

Melting point >300°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR