Ethyl-4-cyano-5-(methylthio)thiophene-2-carboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Cyano-5-methylsulphanylthiophene-2-carboxylic acid ethyl ester, Ethyl 4-cyano-5-(methylthio)thiophene-2-carboxylate
035682-250MG
035682-1G
101839-664EA
92.31
USD
101839-664
101839-666
Ethyl-4-cyano-5-(methylthio)thiophene-2-carboxylate ≥97%
Ethyl-4-cyano-5-(methylthio)thiophene-2-carboxylate
Hazard: Irritant
Formula:
C₉H₉NO₂S₂ MW: 227.31 g/mol Melting Pt: 98…100 °C |
MDL Number:
MFCD01859680 CAS Number: 116170-84-4 |
Specification Test Results
Melting point | 98-100°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...