4-Fluoro-3-(trifluoromethyl)phenylboronic acid ≥95%
Synonyms:
4-Fluoro-3-(trifluoromethyl)benzeneboronic acid
035597-1G
035597-5G
101839-544
101839-546
4-Fluoro-3-(trifluoromethyl)phenylboronic acid ≥95%
4-Fluoro-3-(trifluoromethyl)phenylboronic acid
Hazard: Irritant/Store in Freezer
Formula:
C₇H₅BF₄O₂ MW: 207.92 g/mol Melting Pt: 207…210 °C |
MDL Number:
MFCD07363787 CAS Number: 182344-23-6 |
Specification Test Results
Melting point | 207-210°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...