4-Fluoro-2-methylphenylboronic acid

Supplier: Matrix Scientific

Synonyms: 4-Fluoro-2-methylbenzeneboronic acid, 2-Methyl-4-fluorophenylboronic acid

035595-5G 035595-25G
101839-540EA 52.31 USD
101839-540 101839-542
4-Fluoro-2-methylphenylboronic acid
4-Fluoro-2-methylphenylboronic acid

Hazard: Irritant

Formula: C₇H₈BFO₂
MW: 153.95 g/mol
Melting Pt: 191…196 °C
MDL Number: MFCD02093072
CAS Number: 139911-29-8

Order Now

Specification Test Results

Melting point 191-196°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR