4-Amino-3-chloro-2,5,6-trifluoropyridine ≥98%

Supplier: Matrix Scientific

035279-5G 035279-25G
101839-344EA 207.69 USD
101839-344 101839-346
4-Amino-3-chloro-2,5,6-trifluoropyridine ≥98%
4-Amino-3-chloro-2,5,6-trifluoropyridine

Hazard: Irritant

Formula: C₅H₂ClF₃N₂
MW: 182.53 g/mol
Melting Pt: 120…121 °C
MDL Number: MFCD00010856
CAS Number: 2693-57-4

Order Now

Specification Test Results

Melting point 120-121°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR