2-Amino-5-methylthiophene-3-carbonitrile ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Amino-5-methyl-3-thiophenecarbonitrile, 2-Amino-3-cyano-5-methylthiophene
035278-25G
035278-100G
101839-342EA
130.77
USD
101839-342
102551-170
2-Amino-5-methylthiophene-3-carbonitrile ≥97%
2-Amino-5-methylthiophene-3-carbonitrile
Hazard: Irritant
Formula:
C₆H₆N₂S MW: 138.1902 g/mol Melting Pt: 103…104 °C |
MDL Number:
MFCD01626055 CAS Number: 138564-58-6 |
Specification Test Results
Melting point | 103-104°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...