Ethyl-2-amino-4-methylthiazole-5-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-amino-4-methylthiazole-5-carboxylate

034869-25G 034869-100G
101838-526EA 56.92 USD
101838-526 101838-528
Ethyl-2-amino-4-methylthiazole-5-carboxylate ≥97%
Ethyl 2-amino-4-methylthiazole-5-carboxylate

Hazard: Irritant

Formula: C₇H₁₀N₂O₂S
MW: 186.22 g/mol
Melting Pt: 176-180°C
MDL Number: MFCD00123414
CAS Number: 7210-76-6

Order Now

Specification Test Results

Melting point 176-180°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR