Benzamidoxime ≥97%

Supplier: Matrix Scientific

034856-5G 034856-25G
101838-518EA 60 USD
101838-518 101838-520
Benzamidoxime ≥97%
Benzamidoxime

Hazard: Irritant

Formula: C₇H₈N₂O
Melting Pt: 77°C
MDL Number: MFCD00031485
CAS Number: 613-92-3

Order Now

Specification Test Results

Melting point 77°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR