4-tert-Butylphenylacetic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(4-(tert-Butyl)phenyl)acetic acid

034848-1G 034848-5G
101838-494EA 15.38 USD
101838-494 101838-496
4-tert-Butylphenylacetic acid ≥98%
4-tert-Butylphenylacetic acid

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
Melting Pt: 77…78 °C
MDL Number: MFCD00082593
CAS Number: 32857-63-9

Order Now

Specification Test Results

Melting point 77-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR