2,6-Difluoronicotinic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2,6-Difluoropyridine-3-carboxylic acid

034763-1G 034763-5G 034763-25G
101838-316EA 364.62 USD
101838-316 101838-318 101838-320
2,6-Difluoronicotinic acid ≥97%
2,6-Difluoronicotinic acid

Hazard: Irritant

Formula: C₆H₃F₂NO₂
MW: 159.0900 g/mol
Melting Pt: 165…167 °C
MDL Number: MFCD08064047
CAS Number: 171178-50-0

Order Now

Specification Test Results

Melting point 165-167°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR