3-(Trifluoromethyl)picolinic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Trifluoromethyl)pyridine-2-carboxylic acid, 3-(Trifluoromethy)pyridine-2-carboxylic acid

034757-1G 034757-5G
101838-304EA 26.15 USD
101838-304 101838-306
3-(Trifluoromethyl)picolinic acid ≥97%
3-(Trifluoromethyl)picolinic acid

Hazard: Irritant

Formula: C₇H₄F₃NO₂
MW: 191.1100 g/mol
Melting Pt: 117-118°C
MDL Number: MFCD07375386
CAS Number: 87407-12-3

Order Now

Specification Test Results

Melting point 117-118°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR