Tetrahydropyran-4-carboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: Tetrahydropyran-4-yl-carboxylic acid

034746-5G 034746-25G
101838-274EA 21.54 USD
101838-274 101838-276
Tetrahydropyran-4-carboxylic acid ≥98%
Tetrahydropyran-4-carboxylic acid

Hazard: Irritant

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 146-147°C
Melting Pt: 85-87
MDL Number: MFCD00031016
CAS Number: 5337-03-1

Order Now

Specification Test Results

Melting point 85-87

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR