2-Mercapto-4-(trifluoromethyl)pyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Sulphanyl-4-(trifluoromethyl)pyrimidine

032519-1G 032519-5G
101833-912EA 27.69 USD
101833-912 101833-914
2-Mercapto-4-(trifluoromethyl)pyrimidine ≥95%
2-Mercapto-4-(trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₅H₃F₃N₂S
MW: 180.148 g/mol
Melting Pt: 164…166 °C
MDL Number: MFCD00115113
CAS Number: 136547-17-6

Order Now

Specification Test Results

Melting point 164-166°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR