6-Bromo-1H-indole-2-carboxylic acid ≥98%

Supplier: Matrix Scientific

032436-1G 032436-5G
101833-724EA 21.54 USD
101833-724 101833-726
6-Bromo-1H-indole-2-carboxylic acid ≥98%
6-Bromo-1H-indole-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₆BrNO₂
Melting Pt: 221…223 °C
MDL Number: MFCD02664469
CAS Number: 16732-65-3

Order Now

Specification Test Results

Melting point 221-223°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR