3-Chloro-2-iodo-5-trifluoromethylpyridine

Supplier: Matrix Scientific

024870-250MG 024870-1G
101819-116EA 315.38 USD
101819-116 101819-118
3-Chloro-2-iodo-5-trifluoromethylpyridine
3-Chloro-2-iodo-5-trifluoromethylpyridine

Hazard: Irritant

Formula: C₆H₂ClF₃IN
MW: 307.44 g/mol
Melting Pt: 39…41 °C
MDL Number: MFCD09264543
CAS Number: 134161-12-9

Order Now

Specification Test Results

Melting point 39 - 41 oC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR