3-Fluoro-2-nitrophenol ≥98%

Supplier: Matrix Scientific

024846-5G 024846-25G
101819-058EA 38.46 USD
101819-058 101819-060
3-Fluoro-2-nitrophenol ≥98%
3-Fluoro-2-nitrophenol

Hazard: Irritant

Formula: C₆H₄FNO₃
Melting Pt: 37…38 °C
MDL Number: MFCD07368753
CAS Number: 385-01-3

Order Now

Specification Test Results

Melting point 37-38°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR