(2-Chloro-4-pyridinyl)methanol ≥97%
Supplier: Matrix Scientific
Synonyms:
(2-Chloropyridin-4-yl)methanol, 2-Chloro-4-(hydroxymethyl)pyridine, 2-Chloropyridine-4-methanol
024833-1G
024833-5G
101819-024EA
29.23
USD
101819-024
101819-026
(2-Chloro-4-pyridinyl)methanol ≥97%
(2-Chloro-4-pyridinyl)methanol
Hazard: Irritant
Formula:
C₆H₆ClNO MW: 143.57 g/mol Melting Pt: 64-66°C |
MDL Number:
MFCD06858767 CAS Number: 100704-10-7 |
Specification Test Results
Melting point | 64-66°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...