3-Fluoro-2-methylphenol ≥98%

Supplier: Matrix Scientific

024823-5G 024823-25G
101818-996EA 43.08 USD
101818-996 101818-998
3-Fluoro-2-methylphenol ≥98%
3-Fluoro-2-methylphenol

Hazard: Irritant

Formula: C₇H₇FO
Melting Pt: 56.5 °C
MDL Number: MFCD03788546
CAS Number: 443-87-8

Order Now

Specification Test Results

Melting point 56.5°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR