2,4-Dichloro-5-methoxypyrimidine ≥97%

Supplier: Matrix Scientific

024776-25G 024776-100G
101818-878EA 67.69 USD
101818-878 102551-074
2,4-Dichloro-5-methoxypyrimidine ≥97%
2,4-Dichloro-5-methoxypyrimidine

Hazard: Irritant-Harmful/Lachrymator/Avoid Skin Contact

Formula: C₅H₄Cl₂N₂O
Melting Pt: 66…70 °C
MDL Number: MFCD02685670
CAS Number: 19646-07-2

Order Now

Specification Test Results

Melting point 66-70°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR