4-Fluoro-3-formylphenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Fluoro-3-formylbenzeneboronic acid, (4-Fluoro-3-formylphenyl)boronic acid

024774-1G 024774-5G 024774-25G
101818-868EA 20 USD
101818-868 101818-870 101818-872
4-Fluoro-3-formylphenylboronic acid ≥97%
4-Fluoro-3-formylphenylboronic acid

Hazard: Irritant/Air Sensitive

Formula: C₇H₆BFO₃
MW: 167.93 g/mol
Melting Pt: 132-134°C
MDL Number: MFCD02093074
CAS Number: 374538-01-9

Order Now

Specification Test Results

Melting point 132-134°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR