4-Amino-6-hydroxypyrimidine ≥98%

Supplier: Matrix Scientific

024745-1G 024745-5G
101818-798EA 23.08 USD
101818-798 101818-800
4-Amino-6-hydroxypyrimidine ≥98%
4-Amino-6-hydroxypyrimidine

Hazard: Irritant

Formula: C₄H₅N₃O
MW: 111.10 g/mol
Melting Pt: 262…264 °C
MDL Number: MFCD00051502
CAS Number: 1193-22-2

Order Now

Specification Test Results

Melting point 262-264°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR