3-(Trifluoromethyl)-2-pyridinethiol ≥98%

Supplier: Matrix Scientific

Synonyms: 3-(Trifluoromethyl)pyridine-2-thiol

024710-250MG 024710-1G
101818-706EA 47.69 USD
101818-706 101818-708
3-(Trifluoromethyl)-2-pyridinethiol ≥98%
3-(Trifluoromethyl)-2-pyridinethiol

Hazard: Irritant

Formula: C₆H₄F₃NS
MW: 179.16 g/mol
Melting Pt: 180…182 °C
MDL Number: MFCD00178749
CAS Number: 104040-74-6

Order Now

Specification Test Results

Melting point 180-182°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR