5-Bromonicotinamide ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-3-pyridinecarboxamide

024701-5G 024701-25G 024701-100G
101818-670EA 15.38 USD
101818-670 101818-672 101818-674
5-Bromonicotinamide ≥98%
5-Bromonicotinamide

Hazard: Irritant

Formula: C₆H₅BrN₂O
MW: 201.03 g/mol
Melting Pt: 226…227 °C
MDL Number: MFCD00173919
CAS Number: 28733-43-9

Order Now

Specification Test Results

Melting point 226-227°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR