Ethyl-1,3-benzothiazole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Benzothiazole-2-carboxylic acid ethyl ester, Ethyl 1,3-benzothiazole-2-carboxylate, Ethyl Benzothiazole-2-carboxylate

024700-1G 024700-5G
101818-666EA 15.38 USD
101818-666 101818-668
Ethyl-1,3-benzothiazole-2-carboxylate ≥95%
Ethyl-1,3-benzothiazole-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₉NO₂S
MW: 207.25 g/mol
Melting Pt: 71…72 °C
MDL Number: MFCD00848360
CAS Number: 32137-76-1

Order Now

Specification Test Results

Melting point 71-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR