2,6-Dimethylpyridin-4-ol ≥98%

Supplier: Matrix Scientific

Synonyms: 2,6-Dimethyl-4-pyridinol, 2,6-Dimethyl-4-hydroxypyridine

024641-5G 024641-25G
101818-526EA 24.62 USD
101818-526 101818-528
2,6-Dimethylpyridin-4-ol ≥98%
2,6-Dimethylpyridin-4-ol

Hazard: Irritant

Formula: C₇H₉NO
MW: 123.1525 g/mol
Melting Pt: 230…232 °C
MDL Number: MFCD00956021
CAS Number: 13603-44-6

Order Now

Specification Test Results

Melting point 230-232°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR