Phenylmaleic anhydride ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Phenylfuran-2,5-dione, 3-Phenyl-2,5-furandione

024624-1G 024624-5G
101818-470EA 18.46 USD
101818-470 101818-472
Phenylmaleic anhydride ≥97%
Phenylmaleic anhydride

Hazard: Irritant

Formula: C₁₀H₆O₃
Melting Pt: 118…119 °C
MDL Number: MFCD00015472
CAS Number: 36122-35-7

Order Now

Specification Test Results

Melting point 118-119°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR