2-Chloro-4,6-dimethylnicotinonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-3-cyano-4,6-dimethylpyridine, 2-Chloro-4,6-dimethylpyridine-3-carbonitrile

024597-5G 024597-25G
101818-394EA 64.62 USD
101818-394 101818-396
2-Chloro-4,6-dimethylnicotinonitrile ≥97%
2-Chloro-4,6-dimethylnicotinonitrile

Hazard: Irritant

Formula: C₈H₇ClN₂
MW: 166.61 g/mol
Melting Pt: 97…99 °C
MDL Number: MFCD00051676
CAS Number: 14237-71-9

Order Now

Specification Test Results

Melting point 97-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR