6-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Chloro-2-(trifluoromethyl)quinolin-4-ol

024586-1G 024586-5G
101818-372EA 26.15 USD
101818-372 101818-374
6-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline ≥97%
6-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline

Hazard: Irritant

Formula: C₁₀H₅ClF₃NO
MW: 247.61 g/mol
Melting Pt: 278…280 °C
MDL Number: MFCD00276583
CAS Number: 18706-21-3

Order Now

Specification Test Results

Melting point 275-277°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR