4-(Trifluoromethylsulfonyl)benzonitrile ≥98%

Supplier: Matrix Scientific

024585-1G 024585-5G
101818-368EA 53.85 USD
101818-368 101818-370
4-(Trifluoromethylsulfonyl)benzonitrile ≥98%
4-(Trifluoromethylsulphonyl)benzonitrile

Hazard: Irritant

Formula: C₈H₄F₃NO₂S
Melting Pt: 90…91 °C
MDL Number: MFCD01631631
CAS Number: 312-21-0

Order Now

Specification Test Results

Melting point 90-91°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR