1H-Benzimidazole-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
027197-1G
027197-5G
027197-25G
101823-700EA
53.85
USD
101823-700
101823-702
101823-704
1H-Benzimidazole-2-carboxylic acid ≥95%
1H-Benzimidazole-2-carboxylic acid
Hazard: Irritant
Formula:
C₈H₆N₂O₂ Melting Pt: 165…166 °C |
MDL Number:
MFCD00721565 CAS Number: 2849-93-6 |
Specification Test Results
Melting point | 165-166°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...