3-Chloro-4-methylbenzenesulfonamide ≥97%

Supplier: Matrix Scientific

026764-1G 026764-5G
101822-824EA 63.08 USD
101822-824 101822-826
3-Chloro-4-methylbenzenesulfonamide ≥97%
3-Chloro-4-methylbenzenesulphonamide

Hazard: Irritant

Formula: C₇H₈ClNO₂S
MW: 205.66 g/mol
Melting Pt: 135-138°C
MDL Number: MFCD00025381
CAS Number: 51896-27-6

Order Now

Specification Test Results

Melting point 135-138°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR